Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
1 g | Usually ships in 2 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery) | $529.00 |
About This Item
Empirical Formula (Hill Notation):
C13H13NO5
Molecular Weight:
263.25
UNSPSC Code:
12352106
PubChem Substance ID:
MDL number:
NACRES:
NA.21
Form:
solid
$529.00
Usually ships in 2 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
form
solid
functional group
ketone
SMILES string
O=C1N(C)C2=CC(C(CCC(O)=O)=O)=CC=C2OC1
InChI
1S/C13H13NO5/c1-14-9-6-8(10(15)3-5-13(17)18)2-4-11(9)19-7-12(14)16/h2,4,6H,3,5,7H2,1H3,(H,17,18)
InChI key
AJQGUERUHXEOTE-UHFFFAOYSA-N
Other Notes
Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
1 of 1
This Item | |||
|---|---|---|---|
| description - | description - | description - | description - |
| form solid | form solid | form solid | form solid |
| functional group ketone | functional group carboxylic acid | functional group - | functional group ketone |
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
![4-(3,4-Dihydro-4-methyl-3-oxo-2H-benzo[b][1,4]oxazin-6-yl)-4-oxobutanoic acid](/deepweb/assets/sigmaaldrich/product/structures/117/164/a71ccbe5-afd7-462f-b311-12585d847af7/640/a71ccbe5-afd7-462f-b311-12585d847af7.png)
![3-(6-Chloro-2,2-dimethyl-3-oxo-2H-benzo[b][1,4]oxazin-4(3H)-yl)propanoic acid](/deepweb/assets/sigmaaldrich/product/structures/373/000/f84d94b0-cdda-40cf-8be5-de8eb51abbe9/640/f84d94b0-cdda-40cf-8be5-de8eb51abbe9.png)
![4-Allyl-6-(2-amino-5-methylthiazol-4-yl)-2H-benzo[b][1,4]oxazin-3(4H)-one](/deepweb/assets/sigmaaldrich/product/structures/192/655/42b90708-3eee-44a6-9ad5-8da682fa1251/640/42b90708-3eee-44a6-9ad5-8da682fa1251.png)
![6-(2-Chloropropanoyl)-4-methyl-2H-benzo[b][1,4]oxazin-3(4H)-one](/deepweb/assets/sigmaaldrich/product/structures/134/645/ef0157bd-c78a-434e-a582-53f905273d17/640/ef0157bd-c78a-434e-a582-53f905273d17.png)