Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
1 g | Usually ships in 2 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery) | $474.00 |
About This Item
Empirical Formula (Hill Notation):
C14H9BrN2O3
Molecular Weight:
333.14
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
NACRES:
NA.21
Form:
solid
$474.00
Usually ships in 2 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
form
solid
functional group
bromo, nitro
SMILES string
BrC1=CC=C(C2=NC3=CC([N+]([O-])=O)=CC=C3OC2)C=C1
InChI
1S/C14H9BrN2O3/c15-10-3-1-9(2-4-10)13-8-20-14-6-5-11(17(18)19)7-12(14)16-13/h1-7H,8H2
InChI key
AVHILIMDPZXDIP-UHFFFAOYSA-N
Other Notes
Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY, (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE, OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY, WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY, (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE, OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY, WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
1 of 1
This Item | |||
|---|---|---|---|
| description - | description - | description - | description - |
| form solid | form solid | form solid | form solid |
| functional group nitro, bromo | functional group nitro | functional group fluoro, nitro | functional group nitro |
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
![3-(4-Bromophenyl)-6-nitro-2H-benzo[b][1,4]oxazine](/deepweb/assets/sigmaaldrich/product/structures/148/994/8687839f-9561-466a-b672-46346ee4ebbf/640/8687839f-9561-466a-b672-46346ee4ebbf.png)
![3-(4-Chlorophenyl)-6-nitro-2H-benzo[b][1,4]oxazine](/deepweb/assets/sigmaaldrich/product/structures/995/359/636198b3-d4a0-4080-8e80-3cf954f2777a/640/636198b3-d4a0-4080-8e80-3cf954f2777a.png)
![3-(4-Fluorophenyl)-6-nitro-2H-benzo[b][1,4]oxazine](/deepweb/assets/sigmaaldrich/product/structures/243/942/8d94cc66-bf70-439a-a764-686e39b7b931/640/8d94cc66-bf70-439a-a764-686e39b7b931.png)
![6-Nitro-3,4-dihydro-2H-benzo[b][1,4]oxazine](/deepweb/assets/sigmaaldrich/product/structures/119/329/56672019-b5bc-4a33-be91-c2c7f3d4cbf5/640/56672019-b5bc-4a33-be91-c2c7f3d4cbf5.png)