Skip to Content
MilliporeSigma

Skip To

A-213

Angiotensin I solution

100 μg/mL (Aqueous 10% ACN &0.1% Formic Acid), certified reference material, ampule of 1 mL, Cerilliant®

Synonym(s):

Angiotensin I human acetate salt hydrate, Hypertensin I

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C62H89N17O14 · xC2H4O2 · yH2O
CAS Number:
Molecular Weight:
1296.48 (anhydrous free base basis)
NACRES:
NA.21
MDL number:
UNSPSC Code:
12352202
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL (Aqueous 10% ACN &0.1% Formic Acid)

format

single component solution

storage temp.

-10 to -25°C

SMILES string

CC(O)=O.CC(O)=O.CC[C@H](C)[C@H](NC(=O)[C@H](Cc1ccc(O)cc1)NC(=O)[C@@H](NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@@H](N)CC(O)=O)C(C)C)C(=O)N[C@@H](Cc2c[nH]cn2)C(=O)N3CCC[C@H]3C(=O)N[C@@H](Cc4ccccc4)C(=O)N[C@H](Cc5c[nH]cn5)C(=O)N[C@@H](CC(C)C)C(O)=O

InChI

1S/C62H89N17O14.2C2H4O2/c1-7-35(6)51(78-56(87)44(25-37-17-19-40(80)20-18-37)74-58(89)50(34(4)5)77-53(84)42(15-11-21-68-62(64)65)71-52(83)41(63)28-49(81)82)59(90)75-46(27-39-30-67-32-70-39)60(91)79-22-12-16-48(79)57(88)73-43(24-36-13-9-8-10-14-36)54(85)72-45(26-38-29-66-31-69-38)55(86)76-47(61(92)93)23-33(2)3;2*1-2(3)4/h8-10,13-14,17-20,29-35,41-48,50-51,80H,7,11-12,15-16,21-28,63H2,1-6H3,(H,66,69)(H,67,70)(H,71,83)(H,72,85)(H,73,88)(H,74,89)(H,75,90)(H,76,86)(H,77,84)(H,78,87)(H,81,82)(H,92,93)(H4,64,65,68);2*1H3,(H,3,4)/t35-,41-,42-,43-,44-,45+,46-,47-,48-,50-,51-;;/m0../s1

InChI key

CKIFOTYDIFUKEP-AUVUXLIHSA-N

General description

Asp-Arg-Val-Tyr-Ile-His-Pro-Phe-His-Leu-OH.2CH3COOH.6H2O

Biochem/physiol Actions

Angiotensin I is a putative neurotransmitter. It is the precursor of angiotensin II and of angiotensin fragment 1-7, which are the hormones involved in regulation of fluid volume and the release of aldosterone.
Putative neurotransmitter; precursor of angiotensin II and of angiotensin fragment 1-7.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany


signalword

Warning

Storage Class

3 - Flammable liquids

flash_point_f

113.0 °F

flash_point_c

45 °C

pictograms

FlameExclamation mark

hcodes

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library