Skip to Content
MilliporeSigma

Skip To

8.41569

9-Phenyl-2,3,7-trihydroxy-6-fluorone

for synthesis

Synonym(s):

9-Phenyl-2,3,7-trihydroxy-6-fluorone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 g
Check Cart for Availability
$42.40
5 g
Check Cart for Availability
$140.00

About This Item

Empirical Formula (Hill Notation):
C19H12O5
CAS Number:
Molecular Weight:
320.30
MDL number:
UNSPSC Code:
12352115
EC Index Number:
213-550-3
NACRES:
NA.22
Form:
powder

$42.40


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

powder

mp

>300 °C

storage temp.

2-30°C

SMILES string

[o]1c2c(c(c4c1cc(c(c4)O)O)c3ccccc3)cc([c](c2)=O)O

InChI

1S/C19H12O5/c20-13-6-11-17(8-15(13)22)24-18-9-16(23)14(21)7-12(18)19(11)10-4-2-1-3-5-10/h1-9,20-22H

InChI key

YDCFOUBAMGLLKA-UHFFFAOYSA-N

General description

9-Phenyl-2,3,7-trihydroxy-6-fluorone is commonly used as a precursor for synthesizing dyes and pigments. Additionally, it acts as a ligand in the synthesis of metal-organic frameworks (MOFs) due to the presence of hydroxyl groups.

Analysis Note

Identity (IR): passes test


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters