Skip to Content
MilliporeSigma

Skip To

8.52406

Fmoc-Ser(tBu)-(Dmb)Gly-OH

Novabiochem®

Synonym(s):

Fmoc-Ser(tBu)-(Dmb)Gly-OH

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 g
Check Cart for Availability
$267.00

About This Item

Empirical Formula (Hill Notation):
C33H38N2O8
Molecular Weight:
590.66
NACRES:
NA.22
UNSPSC Code:
12352209

$267.00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Novabiochem®

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

hydroxyl

storage temp.

2-8°C

General description

Fmoc-Aaa-(Dmb)Gly-OH dipeptides offer the same benefits as pseudoproline dipeptides such as prevention of aggregation and improvements in product quality but for peptide sequences containing Gly. They are extremely easy to use. Standard coupling methods like PyBOP®/DIPEA or DIPCDI/HOBt can be used for their introduction. Removal of the Dmb group and regeneration of the glycine residue occurs during the course of standard TFA-mediated cleavage reaction.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Overcoming Aspartimide Formation in Fmoc SPPS

Analysis Note

Colour (visual): white to off white
Appearance of substance (visual): powder
Identity (IR): passes test
Optical rotation α 25/D (c=1 in methanol): -26.0 - -22.0 °
Purity (TLC(157A)): ≥ 98 %
Assay (HPLC, area%): ≥ 97.0 % (a/a)
Solubility (1 mole in 2 ml DMF): clearly soluble
Assay (acidimetric): ≥ 90.0 %
Water (K. F.): ≤ 1.00 %
Ethyl acetate (HS-GC): ≤ 0.5 %
Acetate (IC): ≤ 0.02 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany
PyBOP is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
852405852407852114
description

Novabiochem®

description

Novabiochem®

description

Novabiochem®

description

Novabiochem®

form

powder

form

powder

form

powder

form

powder

functional group

hydroxyl

functional group

carboxylic acid

functional group

amine

functional group

Fmoc

product line

Novabiochem®

product line

Novabiochem®

product line

Novabiochem®

product line

Novabiochem®

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

manufacturer/tradename

Novabiochem®

manufacturer/tradename

Novabiochem®

manufacturer/tradename

Novabiochem®

manufacturer/tradename

Novabiochem®


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters