Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
0.4 mg | Check Cart for Availability | $518.00 |
About This Item
NACRES:
NA.25
UNSPSC Code:
12352201
carrier
PAA carrier
label
FITC labeled
product line
FP-Series
assay
95% (H-NMR)
form
lyophilized powder
mol wt
~20,000 Da
manufacturer/tradename
GlycoNZ
extent of labeling
1% Biotin per 20 mole % Poly[N-(2-hydroxyethyl)acrylamide-co-N-acrylamide] ligand density
technique(s)
flow cytometry: suitable
probe type
trisaccharide
probe presentation
polymeric
storage temp.
−20°C
General description
FP-Series: Fluorescein-Labeled Polymeric Probes
These glycoconjugate probes are designed to reveal lectins, including selectins, directly on cell membranes, organelles, and in cytoplasm. The low fluorescein content ensures that probe solubility in water is not affected, but still allows reliable probe detection. The affinity of the probes is 102 to 105 times higher than that of corresponding free sugars.
PAA is poly[N-(2-hydroxyethyl)acrylamide], a flexible polymer chain which serves as an additional spacer. Used as a molecular weight carrier, PAA demonstrates an absence of non-specific interaction with cells, and is stable to chemical and proteolytic action.
These glycoconjugate probes are designed to reveal lectins, including selectins, directly on cell membranes, organelles, and in cytoplasm. The low fluorescein content ensures that probe solubility in water is not affected, but still allows reliable probe detection. The affinity of the probes is 102 to 105 times higher than that of corresponding free sugars.
PAA is poly[N-(2-hydroxyethyl)acrylamide], a flexible polymer chain which serves as an additional spacer. Used as a molecular weight carrier, PAA demonstrates an absence of non-specific interaction with cells, and is stable to chemical and proteolytic action.
Application
Glycoconjugates are used to study the functions of carbohydrates, carbohydrate-binding proteins in biological processes, and on the discovery of new carbohydrate-mediated interactions.
Preparation Note
Purity: synthetic ω-aminoalkyl glycosides used for coupling with the polymer have a purity of >95% (HPLC and 1H-NMR)
1 of 1
This Item | |||
|---|---|---|---|
| description - | description - | description - | description - |
| carrier PAA carrier | carrier PAA carrier | carrier PAA carrier | carrier PAA carrier |
| probe type trisaccharide | probe type trisaccharide | probe type trisaccharide | probe type tetrasaccharide |
| probe presentation polymeric | probe presentation polymeric | probe presentation polymeric | probe presentation polymeric |
| technique(s) flow cytometry: suitable | technique(s) flow cytometry: suitable | technique(s) flow cytometry: suitable | technique(s) flow cytometry: suitable |
| product line FP-Series | product line FP-Series | product line FP-Series | product line FP-Series |
| assay 95% (H-NMR) | assay 95% (H-NMR) | assay 95% (H-NMR) | assay 95% (H-NMR) |
Storage Class
13 - Non Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
