Skip to Content
MilliporeSigma

Skip To

0088-BP

Bdi-C3-PAA-biot

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mg
Check Cart for Availability
$346.00

About This Item

NACRES:
NA.79
UNSPSC Code:
12352201

$346.00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


carrier

PAA carrier

conjugate

biotin conjugate

product line

BP-Series

assay

95% (H-NMR)

form

lyophilized powder

mol wt

~20,000 Da

manufacturer/tradename

GlycoNZ

extent of labeling

20 mol% Biotin ligand density

technique(s)

flow cytometry: suitable, glycol-bead construction: suitable, glycol-surface construction: suitable

probe type

disaccharide

probe presentation

polymeric

application(s)

histochemistry

storage temp.

−20°C

General description

BP-Series: Biotinylated Polymeric Probes
This series of glycoconjugate probes includes HOCH2CH2NH3+-salts for acidic sugars. The affinity of these probes is 102 to 105 times higher than that of the corresponding free sugars. The spacer-arm is normally –(CH2)3–, while the spacer arm for biotin is –(CH2)6–. The flexible polymer chain behaves as an additional spacer.

Used as a high molecular weight carrier, poly[N-(2-hydroxyethyl)acrylamide], abbreviated as PAA, has low non-specific sorption, and is stable to chemical and proteolytic action.

Application

Glycoconjugates are used to study the functions of carbohydrates, carbohydrate-binding proteins in biological processes, and on the discovery of new carbohydrate-mediated interactions.

Preparation Note

Purity: synthetic ω-aminoalkyl glycosides used for coupling with the polymer have a purity of >95% (HPLC and 1H-NMR)


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters