Skip to Content
MilliporeSigma

Skip To

0745-FP

Fucβ2`(Fucα3)LN-C3-PAA-fluo

Promotion Available

Sign In to View Organizational & Contract Pricing.

Spend $500 — Save 5%; Spend $800 — Save 10%; Spend $1,600 — Save 22%*

Sign in to check eligibility. Shop Products

Select a Size

Change View
Size/SKUAvailabilityPrice
0.4 mg
Check Cart for Availability
$864.00

About This Item

NACRES:
NA.25
UNSPSC Code:
12352201

$864.00

Save Up To 22%


Check Cart for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


carrier

PAA carrier

label

FITC labeled

product line

FP-Series

assay

95% (H-NMR)

form

lyophilized powder

mol wt

~20,000 Da

manufacturer/tradename

GlycoNZ

extent of labeling

1% Biotin per 20 mole % Poly[N-(2-hydroxyethyl)acrylamide-co-N-acrylamide] ligand density

technique(s)

flow cytometry: suitable

probe type

tetrasaccharide

probe presentation

polymeric

storage temp.

−20°C

General description

FP-Series: Fluorescein-Labeled Polymeric Probes
These glycoconjugate probes are designed to reveal lectins, including selectins, directly on cell membranes, organelles, and in cytoplasm. The low fluorescein content ensures that probe solubility in water is not affected, but still allows reliable probe detection. The affinity of the probes is 102 to 105 times higher than that of corresponding free sugars.

PAA is poly[N-(2-hydroxyethyl)acrylamide], a flexible polymer chain which serves as an additional spacer. Used as a molecular weight carrier, PAA demonstrates an absence of non-specific interaction with cells, and is stable to chemical and proteolytic action.

Application

Glycoconjugates are used to study the functions of carbohydrates, carbohydrate-binding proteins in biological processes, and on the discovery of new carbohydrate-mediated interactions.

Preparation Note

Purity: synthetic ω-aminoalkyl glycosides used for coupling with the polymer have a purity of >95% (HPLC and 1H-NMR)


Storage Class

13 - Non Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters