Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 mg | Check Cart for Availability | $518.00 |
About This Item
conjugate
biotin conjugate
product line
BM-Series
assay
95% (H-NMR)
form
lyophilized powder
mol wt
983.1 Da
manufacturer/tradename
GlycoNZ
extent of labeling
20 mol% Biotin ligand density
technique(s)
glycol-bead construction: suitable, glycol-surface construction: suitable, microarray: suitable
probe type
trisaccharide
probe presentation
monomeric
storage temp.
−20°C
General description
These glycoconjugate probes have an elongated spacer giving them the possibility to bind avidin or streptavidin. The biotin fragment is hydrophobic enough to permit the probes to be adsorbed on SepPack C18 cartridges from aqueous solutions, and to be eluted by methanol, similarly to glycosides bearing a “Lemieux-type” space. The spacer is the normal spacer of the ligand plus the spacer of the biotin label –CO(CH2)5NH–.
Application
1 of 1
This Item | |||
|---|---|---|---|
| description - | description - | description - | description - |
| probe type trisaccharide | probe type trisaccharide | probe type disaccharide | probe type disaccharide |
| probe presentation monomeric | probe presentation monomeric | probe presentation monomeric | probe presentation monomeric |
| conjugate biotin conjugate | conjugate biotin conjugate | conjugate biotin conjugate | conjugate biotin conjugate |
| technique(s) glycol-bead construction: suitable, microarray: suitable, glycol-surface construction: suitable | technique(s) glycol-bead construction: suitable, glycol-surface construction: suitable, microarray: suitable | technique(s) glycol-bead construction: suitable, glycol-surface construction: suitable, microarray: suitable | technique(s) glycol-bead construction: suitable, glycol-surface construction: suitable, microarray: suitable |
| product line BM-Series | product line BM-Series | product line BM-Series | product line BM-Series |
| assay 95% (H-NMR) | assay 95% (H-NMR) | assay 95% (H-NMR) | assay 95% (H-NMR) |
Storage Class
13 - Non Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
