Skip to Content
MilliporeSigma

Skip To

0809-PA

(GN)3-3,4,6GalNAcα-C3-PAA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mg
Check Cart for Availability
$648.00

About This Item

NACRES:
NA.25
UNSPSC Code:
12352201

$648.00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


carrier

PAA carrier

product line

PA-Series

assay

95% (H-NMR)

form

lyophilized powder

mol wt

~20,000 Da

manufacturer/tradename

GlycoNZ

extent of labeling

20 mol% Poly[N-(2-hydroxyethyl)acrylamide-co-N-acrylamide] ligand density

technique(s)

ELISA: suitable

probe type

tetrasaccharide

probe presentation

polymeric

storage temp.

−20°C

General description

PA-Series: Unlabeled Polyacrylamide Conjugates
This series of glycoconjugate probes consists of HOCH2CH2NH3+-salts for carboxylates and sulfates. PAA is poly[N-(2-hydroxyethyl) acrylamide], a flexible polymer chain which serves as an additional spacer. The spacer-arm for oligosaccharides is normally –(CH2)3–.

Application

Glycoconjugates are used to study the functions of carbohydrates, carbohydrate-binding proteins in biological processes, and on the discovery of new carbohydrate-mediated interactions.

Preparation Note

Purity: synthetic ω-aminoalkyl glycosides used for coupling with the polymer have a purity of >95% (HPLC and 1H-NMR)


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters