Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 mg | Check Cart for Availability | $346.00 |
1 mg | Check Cart for Availability | $346.00 |
About This Item
carrier
PAA carrier
conjugate
biotin conjugate
product line
BP-Series
assay
95% (H-NMR)
form
lyophilized powder
mol wt
~20,000 Da
manufacturer/tradename
GlycoNZ
extent of labeling
20 mol% Biotin ligand density
technique(s)
flow cytometry: suitable, glycol-bead construction: suitable, glycol-surface construction: suitable
probe type
disaccharide
probe presentation
polymeric
application(s)
histochemistry
storage temp.
−20°C
General description
This series of glycoconjugate probes includes HOCH2CH2NH3+-salts for acidic sugars. The affinity of these probes is 102 to 105 times higher than that of the corresponding free sugars. The spacer-arm is normally –(CH2)3–, while the spacer arm for biotin is –(CH2)6–. The flexible polymer chain behaves as an additional spacer.
Used as a high molecular weight carrier, poly[N-(2-hydroxyethyl)acrylamide], abbreviated as PAA, has
low non-specific sorption, and is stable to chemical and proteolytic action.
Application
Preparation Note
1 of 1
This Item | |||
|---|---|---|---|
| description - | description - | description - | description - |
| probe presentation polymeric | probe presentation polymeric | probe presentation polymeric | probe presentation polymeric |
| probe type disaccharide | probe type disaccharide | probe type disaccharide | probe type disaccharide |
| carrier PAA carrier | carrier PAA carrier | carrier PAA carrier | carrier PAA carrier |
| conjugate biotin conjugate | conjugate biotin conjugate | conjugate biotin conjugate | conjugate biotin conjugate |
| technique(s) flow cytometry: suitable, glycol-surface construction: suitable, glycol-bead construction: suitable | technique(s) flow cytometry: suitable, glycol-bead construction: suitable, glycol-surface construction: suitable | technique(s) flow cytometry: suitable, glycol-bead construction: suitable, glycol-surface construction: suitable | technique(s) flow cytometry: suitable, glycol-bead construction: suitable, glycol-surface construction: suitable |
| product line BP-Series | product line BP-Series | product line BP-Series | product line BP-Series |
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
