Skip to Content
MilliporeSigma

Skip To

0871-FP

LN6`LN-C2-PAA-fluo

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
0.4 mg
Check Cart for Availability
$864.00

About This Item

NACRES:
NA.25
UNSPSC Code:
12352201

$864.00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


carrier

PAA carrier

label

FITC labeled

product line

FP-Series

assay

95% (H-NMR)

form

lyophilized powder

mol wt

~20,000 Da

manufacturer/tradename

GlycoNZ

extent of labeling

1% Biotin per 20 mole % Poly[N-(2-hydroxyethyl)acrylamide-co-N-acrylamide] ligand density

technique(s)

flow cytometry: suitable

probe type

tetrasaccharide

probe presentation

polymeric

storage temp.

−20°C

General description

FP-Series: Fluorescein-Labeled Polymeric Probes
These glycoconjugate probes are designed to reveal lectins, including selectins, directly on cell membranes, organelles, and in cytoplasm. The low fluorescein content ensures that probe solubility in water is not affected, but still allows reliable probe detection. The affinity of the probes is 102 to 105 times higher than that of corresponding free sugars.

PAA is poly[N-(2-hydroxyethyl)acrylamide], a flexible polymer chain which serves as an additional spacer. Used as a molecular weight carrier, PAA demonstrates an absence of non-specific interaction with cells, and is stable to chemical and proteolytic action.

Application

Glycoconjugates are used to study the functions of carbohydrates, carbohydrate-binding proteins in biological processes, and on the discovery of new carbohydrate-mediated interactions.

Preparation Note

Purity: synthetic ω-aminoalkyl glycosides used for coupling with the polymer have a purity of >95% (HPLC and 1H-NMR)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
0842FP0867FP0866FP
description

-

description

-

description

-

description

-

probe type

tetrasaccharide

probe type

tetrasaccharide

probe type

hexasaccharide

probe type

pentasaccharide

carrier

PAA carrier

carrier

PAA carrier

carrier

PAA carrier

carrier

PAA carrier

probe presentation

polymeric

probe presentation

polymeric

probe presentation

polymeric

probe presentation

polymeric

technique(s)

flow cytometry: suitable

technique(s)

flow cytometry: suitable

technique(s)

flow cytometry: suitable

technique(s)

flow cytometry: suitable

product line

FP-Series

product line

FP-Series

product line

FP-Series

product line

FP-Series

manufacturer/tradename

GlycoNZ

manufacturer/tradename

GlycoNZ

manufacturer/tradename

GlycoNZ

manufacturer/tradename

GlycoNZ


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters