Skip to Content
MilliporeSigma

Skip To

0884-BM

M5β-Gly-biot

Promotion Available

Sign In to View Organizational & Contract Pricing.

Spend $500 — Save 5%; Spend $800 — Save 10%; Spend $1,600 — Save 22%*

Sign in to check eligibility. Shop Products

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mg

Estimated to ship onOctober 05, 2026fromSAINT LOUIS

$1,300.00

About This Item

NACRES:
NA.79
UNSPSC Code:
12352201

$1,300.00

Save Up To 22%


Estimated to ship onOctober 05, 2026Details


Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


conjugate

biotin conjugate

product line

BM-Series

assay

95% (H-NMR)

form

lyophilized powder

mol wt

1224.2 Da

manufacturer/tradename

GlycoNZ

extent of labeling

20 mol% Biotin ligand density

technique(s)

glycol-bead construction: suitable, glycol-surface construction: suitable, microarray: suitable

probe type

pentasaccharide

probe presentation

monomeric

storage temp.

−20°C

General description

BM-Series: Biotinylated Monomeric Probes
These glycoconjugate probes have an elongated spacer giving them the possibility to bind avidin or streptavidin. The biotin fragment is hydrophobic enough to permit the probes to be adsorbed on SepPack C18 cartridges from aqueous solutions, and to be eluted by methanol, similarly to glycosides bearing a “Lemieux-type” space. The spacer is the normal spacer of the ligand plus the spacer of the biotin label –CO(CH2)5NH–.

Application

Glycoconjugates are used to study the functions of carbohydrates, carbohydrate-binding proteins in biological processes, and on the discovery of new carbohydrate-mediated interactions.


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters