Skip to Content
MilliporeSigma

Skip To

0959-BM

(Glcα6)3β-Gly-biot

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mg
Check Cart for Availability
$432.00

About This Item

NACRES:
NA.25
UNSPSC Code:
12352201

$432.00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


conjugate

biotin conjugate

product line

BM-Series

assay

95% (H-NMR)

form

lyophilized powder

mol wt

899.96 Da

manufacturer/tradename

GlycoNZ

extent of labeling

20 mol% Biotin ligand density

technique(s)

glycol-bead construction: suitable, glycol-surface construction: suitable, microarray: suitable

probe type

trisaccharide

probe presentation

monomeric

storage temp.

−20°C

General description

BM-Series: Biotinylated Monomeric Probes
These glycoconjugate probes have an elongated spacer giving them the possibility to bind avidin or streptavidin. The biotin fragment is hydrophobic enough to permit the probes to be adsorbed on SepPack C18 cartridges from aqueous solutions, and to be eluted by methanol, similarly to glycosides bearing a “Lemieux-type” space. The spacer is the normal spacer of the ligand plus the spacer of the biotin label –CO(CH2)5NH–.

Application

Glycoconjugates are used to study the functions of carbohydrates, carbohydrate-binding proteins in biological processes, and on the discovery of new carbohydrate-mediated interactions.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
0932BM0931BM0609BM
description

-

description

-

description

-

description

-

probe presentation

monomeric

probe presentation

monomeric

probe presentation

monomeric

probe presentation

monomeric

probe type

trisaccharide

probe type

trisaccharide

probe type

tetrasaccharide

probe type

monosaccharide

conjugate

biotin conjugate

conjugate

biotin conjugate

conjugate

biotin conjugate

conjugate

biotin conjugate

technique(s)

glycol-bead construction: suitable, microarray: suitable, glycol-surface construction: suitable

technique(s)

glycol-bead construction: suitable, glycol-surface construction: suitable, microarray: suitable

technique(s)

glycol-bead construction: suitable, glycol-surface construction: suitable, microarray: suitable

technique(s)

glycol-bead construction: suitable, glycol-surface construction: suitable, microarray: suitable

product line

BM-Series

product line

BM-Series

product line

BM-Series

product line

BM-Series

extent of labeling

20 mol% Biotin ligand density

extent of labeling

20 mol% Biotin ligand density

extent of labeling

20 mol% Biotin ligand density

extent of labeling

20 mol% Biotin ligand density


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters