Skip to Content
MilliporeSigma

Skip To

0959-BP

(Glcα6)3β-Gly-PAA-biot

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mg
Check Cart for Availability
$432.00

About This Item

NACRES:
NA.25
UNSPSC Code:
12352201

$432.00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


carrier

PAA carrier

conjugate

biotin conjugate

product line

BP-Series

assay

95% (H-NMR)

form

lyophilized powder

mol wt

~20,000 Da

manufacturer/tradename

GlycoNZ

extent of labeling

20 mol% Biotin ligand density

technique(s)

flow cytometry: suitable, glycol-bead construction: suitable, glycol-surface construction: suitable

probe type

trisaccharide

probe presentation

polymeric

application(s)

histochemistry

storage temp.

−20°C

General description

BP-Series: Biotinylated Polymeric Probes
This series of glycoconjugate probes includes HOCH2CH2NH3+-salts for acidic sugars. The affinity of these probes is 102 to 105 times higher than that of the corresponding free sugars. The spacer-arm is normally –(CH2)3–, while the spacer arm for biotin is –(CH2)6–. The flexible polymer chain behaves as an additional spacer.

Used as a high molecular weight carrier, poly[N-(2-hydroxyethyl)acrylamide], abbreviated as PAA, has
low non-specific sorption, and is stable to chemical and proteolytic action.

Application

Glycoconjugates are used to study the functions of carbohydrates, carbohydrate-binding proteins in biological processes, and on the discovery of new carbohydrate-mediated interactions.

Preparation Note

Purity: synthetic ω-aminoalkyl glycosides used for coupling with the polymer have a purity of >95% (HPLC and 1H-NMR)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
0932BP0931BP0609BP
description

-

description

-

description

-

description

-

conjugate

biotin conjugate

conjugate

biotin conjugate

conjugate

biotin conjugate

conjugate

biotin conjugate

probe type

trisaccharide

probe type

trisaccharide

probe type

tetrasaccharide

probe type

monosaccharide

carrier

PAA carrier

carrier

PAA carrier

carrier

PAA carrier

carrier

PAA carrier

probe presentation

polymeric

probe presentation

polymeric

probe presentation

polymeric

probe presentation

polymeric

technique(s)

flow cytometry: suitable, glycol-surface construction: suitable, glycol-bead construction: suitable

technique(s)

flow cytometry: suitable, glycol-bead construction: suitable, glycol-surface construction: suitable

technique(s)

flow cytometry: suitable, glycol-bead construction: suitable, glycol-surface construction: suitable

technique(s)

flow cytometry: suitable, glycol-bead construction: suitable, glycol-surface construction: suitable

product line

BP-Series

product line

BP-Series

product line

BP-Series

product line

BP-Series


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters