Skip to Content
MilliporeSigma

Skip To

48934-U

ProClin™ 300

pkg of 5 mL

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 pkg

Available to ship TODAY

$40.30

About This Item

UNSPSC Code:
12352200
NACRES:
NB.34
eCl@ss:
32150313

$40.30


Available to ship TODAYDetails


​
​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


shelf life

3 yr (from Date of Manufacture)

Quality Segment

packaging

pkg of 5 mL

Legal Information

ProClin is a trademark of LANXESS Corp.

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada′s Pest Management Regulatory Agency, Turkey′s Biocidal Products Regulation, Korea′s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
48911U48912U48914U
description

pkg of 5 mL

description

ProClin™ preservative for use in IVD manufacturing, CMIT/MIT 3%, Method Development Kit, pkg of 3 × 5 mL each (three different lots)

description

pkg of 50 mL, ProClin™ preservative for use in IVD manufacturing, CMIT/MIT 3 %

description

pkg of 400 mL, ProClin™ preservative for use in IVD manufacturing, CMIT/MIT 3 %

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300

packaging

pkg of 5 mL

packaging

pkg of 3 × 5 mL each (three different lots)

packaging

pkg of 50 mL

packaging

pkg of 400 mL

shelf life

3 yr (from Date of Manufacture)

shelf life

3 yr (from Date of Manufacture)

shelf life

3 yr (from Date of Manufacture)

shelf life

3 yr (from Date of Manufacture)


Still not finding the right product?

Explore all of our products under ProClin™ 300


Signal Word

Danger

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

supp_hazards

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1C - Skin Sens. 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library