Skip to Content
MilliporeSigma

82006BC

Assurance® GDS Adaptor Box Power Supply

Assurance GDS®

Synonym(s):

Power Supply Box

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.61
UNSPSC Code:
41106401
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

Assurance GDS®

technique(s)

PCR: suitable

application(s)

food and beverages

compatibility

for use with BioControl GDS

storage temp.

room temp

General description

The Assurance® GDS Adaptor Box Power Supply is an essential part to be used with the GDS Adaptor Box for the GDS Rotor-Gene Q. As part of the GDS system, the Adaptor Box improves laptop connectivity and PCR run flexibility, allowing one laptop to control up to five Rotor-Genes at one time.

Application

The Assurance® GDS Adaptor Box Power Supply is an essential part used with the Assurance"] GDS Adaptor Box.

Legal Information

Assurance is a registered trademark of Merck KGaA, Darmstadt, Germany
Assurance GDS is a registered trademark of BioControl Systems, Inc.




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library