Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.24
Pricing and availability is not currently available.
grade
certified reference material
agency
(National Research Council of Canada (NRC))
manufacturer/tradename
National Research Council of Canada (NRC)
application(s)
cosmetics
food and beverages
format
matrix material
storage temp.
−20°C (−15°C to −25°C)
General description
CRM-Zero-Mus is a mussel tissue (Mytilus edulis) matrix CRM intended for use as a negative control for the analysis of shellfish toxins. It is free of the following toxins: paralytic shellfish toxins (PSTs), domoic acid (DA) and its isomers, okadaic acid (OA) and dinophysistoxins (DTXs), azaspiracids (AZAs), yessotoxins (YTXs), pectenotoxins (PTXs) and gymnodimine (GYM). It is suited for use in testing analytical procedures for selectivity, spike recovery and matrix matched calibration. CRM-Zero-Mus is not suitable as a negative control for spirolides (SPXs) and pinnatoxins (PnTXs) because these toxins are present, albeit at very low levels.
Features and Benefits
NRC Canada’s certified reference materials (CRMs) are traceable to the International System of Units, and conform to the requirements of ISO/IEC 17025 and ISO 17034.For specific values, as well as additional information regarding stability and storage information, please view the Certificate of Analysis.With NRC Canada’s CRMs, you gain access to products backed by extensive research and expertise, fostering trust and consistency in your results. Elevate your laboratory’s accuracy and credibility with NRC Canada’s certified reference materials.
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Other Notes
Tested for the analytes listed below. See certificate for values and more details
Toxins: saxitoxin, neosaxitoxin, decarbamoylsaxitoxin, gonyautoxin-4, gonyautoxin-1, gonyautoxin-5, decarbamoylgonyautoxin-3, decarbamoylgonyautoxin-2, gonyautoxin-2, gonyautoxin-3, N-Sulfocarbamoylgonyautoxin-2, N-Sulfocarbamoylgonyautoxin-3, N-Sulfocarbamoylgonyautoxin-1, N-Sulfocarbamoylgonyautoxin-4, decarbamoylgonyautoxin-4, decarbamoylgonyautoxin-1, decarbamoylneosaxitoxin, gonyautoxin-6, domoic acid, C5′-epidomoic acid, isodomoic acids A-H, okadaic acid, dinophysistoxin-1, dinophysistoxin-2, azaspiracid-1, azaspiracid-2, azaspiracid-3, yessotoxin, pectenotoxin-2, gymnodimine, 13-desmethyl-spirolide C, 13-desmethyl-spirolide D, pinnatoxin G
Matrix Group: Fish and Seafood
Toxins: saxitoxin, neosaxitoxin, decarbamoylsaxitoxin, gonyautoxin-4, gonyautoxin-1, gonyautoxin-5, decarbamoylgonyautoxin-3, decarbamoylgonyautoxin-2, gonyautoxin-2, gonyautoxin-3, N-Sulfocarbamoylgonyautoxin-2, N-Sulfocarbamoylgonyautoxin-3, N-Sulfocarbamoylgonyautoxin-1, N-Sulfocarbamoylgonyautoxin-4, decarbamoylgonyautoxin-4, decarbamoylgonyautoxin-1, decarbamoylneosaxitoxin, gonyautoxin-6, domoic acid, C5′-epidomoic acid, isodomoic acids A-H, okadaic acid, dinophysistoxin-1, dinophysistoxin-2, azaspiracid-1, azaspiracid-2, azaspiracid-3, yessotoxin, pectenotoxin-2, gymnodimine, 13-desmethyl-spirolide C, 13-desmethyl-spirolide D, pinnatoxin G
Matrix Group: Fish and Seafood
For more information, please refer to the Certificate of Analysis or Safety Data Sheet
Sales restrictions may apply.
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.