Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
50 mg | Estimated to ship onSeptember 14, 2026fromMILWAUKEE | $329.00 $246.75 |
About This Item
$246.75
List Price$329.00Save 25%Estimated to ship onSeptember 14, 2026Details
grade
certified reference material
Quality Segment
packaging
pkg of 50 mg
application(s)
pharmaceutical
format
neat
storage temp.
-10 to -25°C
General description
Application
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| description Pharmaceutical Secondary Standard; Certified Reference Material | description Pharmaceutical Secondary Standard; Certified Reference Material | description certified reference material, pharmaceutical secondary standard | description Pharmaceutical Secondary Standard; Certified Reference Material |
| application(s) pharmaceutical | application(s) pharmaceutical | application(s) pharmaceutical small molecule | application(s) pharmaceutical (small molecule) |
| format neat | format neat | format - | format - |
| Quality Level 300 | Quality Level 300 | Quality Level 300 | Quality Level 300 |
| grade certified reference material | grade certified reference material, pharmaceutical secondary standard | grade pharmaceutical secondary standard, certified reference material | grade certified reference material, pharmaceutical secondary standard |
| storage temp. -10 to -25°C | storage temp. -10 to -25°C | storage temp. 2-30°C | storage temp. 2-30°C |
| packaging pkg of 50 mg | packaging pkg of 100 mg | packaging pkg of 50 mg | packaging ampule of 3x1.2 mL |
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


![2-[4-(2-Hydroxy-2-methylpropyl)-phenyl]propanoic Acid certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/structures/133/400/476867f7-e388-43a9-97a1-8b0458a8e4e3/640/476867f7-e388-43a9-97a1-8b0458a8e4e3.png)
