Skip to Content
MilliporeSigma

Skip To

AV46307

Anti-PPCDC antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-FLJ14585, Anti-MDS018, Anti-Phosphopantothenoylcysteine decarboxylase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μL
Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
$522.00

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
1

$522.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

22 kDa

species reactivity

dog, human, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... PPCDC(60490)

Immunogen

Synthetic peptide directed towards the N terminal region of human PPCDC

Application

Anti-PPCDC antibody produced in rabbit is suitable for western blotting at a concentration of 2.5μg/ml.

Biochem/physiol Actions

PPCDC (phosphopantothenoylcysteine decarboxylase) gene also referred to as FLJ14585 or MDS018 encodes for an enzyme that belongs to larger family of cysteine decarboxylases including the lantibiotic-biosynthesizing enzymes EpiD and MrsD. It catalyzes the biosynthesis of coenzyme A (CoA) from pantothenic acid (vitamin B5). PPCDC is required as one of the last enzymes in CoA synthesis, where it causes decarboxylation of the cysteine moiety of 4′-phosphopantothenoylcysteine (PPC) to form 4′-phosphopantetheine (PPantSH).

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Other Notes

Synthetic peptide located within the following region: VTTERAKHFYSPQDIPVTLYSDADEWEIWKSRSDPVLHIDLRRWADLLLV

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB1407835HPA045667HPA031361
description

IgG fraction of antiserum

description

purified immunoglobulin, buffered aqueous solution

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

species reactivity

dog, human, rat

species reactivity

human

species reactivity

human

species reactivity

human

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

biological source

rabbit

biological source

mouse

biological source

rabbit

biological source

rabbit

antibody form

IgG fraction of antiserum

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

form

buffered aqueous solution

form

buffered aqueous solution

form

buffered aqueous glycerol solution

form

buffered aqueous glycerol solution


Still not finding the right product?

— or —

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library