Skip to Content
MilliporeSigma

AXYMRRB

Corning® Empty Rack for Axygen® MultiRack for 200uL extended length, 300uL extended length, and 1000 uL extended length

5 racks

Synonym(s):

pipet tip, pipette tip, pipettor tips

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41121600
NACRES:
NB.12
Manufacturer/tradename:
Corning MR-R-B
Packaging:
pack of 5 racks
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


manufacturer/tradename

Corning MR-R-B

packaging

pack of 5 racks

General description

Axygen® universal pipet tips are designed to fit a wide variety ofsingle- and multi-channel pipettors. Flexible walls and a series ofinternal sealing rings ensure a secure fit with less force requiredto load and eject.The Axygen product offering now also includes the AxygenMultiRack System, which is our updated reload and rackingsystem. In addition to the universal fit tips packaged in theMultiRack System, we now also offer LTS Compatible tips in theMultiRack System, and if you are worried about your carbonfootprint, the Axygen HybridRack™ is a perfect option for you.

Application

liquid handling

Features and Benefits

  • Automated assembly – Ensures product is free from contamination.
  • ISO 13485 and ISO 14001 – Certified quality management systems supply stringent quality processes to ensure the highest quality finished good.
  • In-house testing – In-house QA lab allows product to be validated for fit, form, and function prior to release.
  • Brand you can trust – A long track record of innovation and value as one of the Corning trusted brands.
  • Broad product offering – Corning offers many different styles of pipet tips, including racked, filtered, reloads, stack racks, low retention, wide bore, and more. With so many options, you are sure to find a tip that works for you.

Legal Information

Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
HybridRack is a trademark of Corning, Inc.




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library