Skip to Content
MilliporeSigma

Skip To

CLS4637

Corning® Spheroid Microplate

1536 well, Black with Clear Round bottom, with Lid, ULA, Sterile, individually wrapped

Synonym(s):

MTP plate, assay plate, microtiter plate, microwell plate, multiwell plate, well plate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41113036
eCl@ss:
32040503
Material:
polystyrene
Sterility:
sterile
Binding type:
low binding surface
Feature:
wells 1536, lid
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polystyrene

sterility

sterile

feature

wells 1536, lid

packaging

case of 5

manufacturer/tradename

Corning 4637

plate wells

14 mL

plate working volume

5 μL (10 μL)

binding type

low binding surface

General description

With their novel and proprietary design, these microplates are ideal for generating and analyzing 3D multicellular spheroids in the same microplate. The Ultra-Low Attachment (ULA) surface enables uniform and reproducible 3D multicellular spheroid formation. The black opaque microplate body shields each optically clear, round bottom well from well-to-well cross-talk.

Application

3D cell culture plate

Features and Benefits

  • 1536-well format allows for high throughput 3D cell culture
  • Optically clear round bottom with black opaque microplate body
  • Covalent attachment of Ultra-Low Attachment surface to reduce cellular adhesion to well surface
  • Novel well geometry aids in the generation of uniform, single spheroids across all wells, which enables automated visualization. Unique design shields each well to minimize well-to-well cross-talk.
  • You can culture and assay spheroids in the same microplate, without the need for transfer to a new microplate.
  • Compatible with the GNF ultra high throughput system

Legal Information

Corning is a registered trademark of Corning, Inc.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
CLS4515CLS4516CLS4527
description

1536 well, Black with Clear Round bottom, with Lid, ULA, Sterile, individually wrapped

description

96 well, round black/clear bottom, ULA (Ultra-Low Attachment surface), lid, sterile

description

384 well, round black/clear bottom, ULA (Ultra-Low Attachment surface), lid, sterile

description

wells (1536), round black/clear bottom, ULA (Ultra-Low Attachment surface), sterile

sterility

sterile

sterility

sterile

sterility

sterile

sterility

sterile

binding type

low binding surface

binding type

(Ultra-Low Attachment)

binding type

(ultra-low attachment)

binding type

-

manufacturer/tradename

Corning 4637

manufacturer/tradename

Corning 4515

manufacturer/tradename

Corning 4516

manufacturer/tradename

Corning 4527

feature

lid, wells 1536

feature

lid, Specific plates for 3D cell culture, skirt, ULA (Ultra-Low Attachment surface)

feature

ULA (Ultra-Low Attachment surface), lid, skirt, Specific plates for 3D cell culture

feature

ULA (Ultra-Low Attachment surface), Specific plates for 3D cell culture, Recommended Working Volume, Lid Included, Recommended Medium Well Volume

material

polystyrene

material

polystyrene , clear bottom, round black/clear bottom

material

polystyrene , round black/clear bottom

material

polystyrene , round black/clear bottom

packaging

case of 5

packaging

pkg of 5 ea

packaging

pkg of 5 ea

packaging

-




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library