Skip to Content
MilliporeSigma

Skip To

CLS8504

Corning® Screw Cap for 96 Storage Tubes

Polypropylene, Orange, with O-Ring, Nonsterile, Bulk, 4800 caps per case

Synonym(s):

sample tube cap, storage tube cap, testing tube screw cap

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
96 each
Check Cart for Availability
$83.80
4800 each
Check Cart for Availability
$3,240.00

About This Item

UNSPSC Code:
41121710
Material:
orange polypropylene
Sterility:
non-sterile
Packaging:
pkg of 4800 per case, 50 packs of 96

$83.80


Check Cart for Availability

​
​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

orange polypropylene

sterility

non-sterile

manufacturer/tradename

Corning 8504

packaging

pkg of 4800 per case, 50 packs of 96

General description

96 Storage Tube Screw Cap, Thermoplastic Elastomer (TPE), Bulk Pack

Application

Sample storage

Features and Benefits

For use with Cat. Nos. 8500 and 4501

Legal Information

Corning is a registered trademark of Corning, Inc.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
AXYST45SCLS8505AXYSCT200SSCS
description

Polypropylene, Orange, with O-Ring, Nonsterile, Bulk, 4800 caps per case

description

capacity 0.5 mL, clear, sterile

description

polypropylene, with o-ring on mat, clear with cover

description

self-standing, size 2.0 mL, clear, pkg of 8x500caps/cs, sterile

material

orange polypropylene

material

screw top cap, clear

material

polypropylene

material

screw top cap, polypropylene , conical bottom tube

packaging

pkg of 4800 per case, 50 packs of 96

packaging

case of 8 packs, pack of 500 tubes

packaging

-

packaging

pkg of 8x500caps/cs

manufacturer/tradename

Corning 8504

manufacturer/tradename

Corning, Axygen Inc

manufacturer/tradename

Corning

manufacturer/tradename

Axygen Inc SCT-200-SS-C-S, Corning

sterility

non-sterile

sterility

sterile

sterility

non-sterile

sterility

sterile




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library