Skip to Content
MilliporeSigma

Skip To

P5867

Anti-Protein 4-Peroxidase Conjugate antibody produced in goat

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
500 μL
Check Cart for Availability
$398.00

About This Item

NACRES:
NA.45
UNSPSC Code:
12352203
Clone:
polyclonal
Species reactivity:
-
Application:
—
Citations:
-

$398.00


Check Cart for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

biological source

goat

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

shipped in

wet ice

storage temp.

−20°C

Application

custom product only

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P5492P5617A4541
description

-

description

-

description

-

description

affinity isolated antibody, buffered aqueous solution

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

antibody form

IgG fraction of antiserum

antibody form

IgG fraction of antiserum

antibody form

IgG fraction of antiserum

antibody form

affinity isolated antibody

biological source

goat

biological source

goat

biological source

goat

biological source

goat

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

dry ice

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

10 - Combustible liquids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library