Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
100 μL | Check Cart for Availability | $540.00 |
About This Item
$540.00
biological source
goat
Quality Segment
conjugate
unconjugated
antibody form
affinity purified immunoglobulin
antibody product type
primary antibodies
clone
polyclonal
species reactivity
human
technique(s)
immunocytochemistry: 1:500-1:2,000, western blot: 1:1,000- 1:10,000
accession no.
NP_005853.2
shipped in
wet ice
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
goat ... SA1(10274)
Immunogen
Physical form
Other Notes
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| description Powered by Bethyl Laboratories, Inc. | description Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution | description affinity isolated antibody | description Powered by Bethyl Laboratories, Inc. |
| antibody form affinity purified immunoglobulin | antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form affinity purified immunoglobulin |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| biological source goat | biological source rabbit | biological source rabbit | biological source rabbit |
| clone polyclonal | clone polyclonal | clone polyclonal | clone polyclonal |
| species reactivity human | species reactivity human, mouse, rat | species reactivity human | species reactivity human, mouse |
| technique(s) western blot: 1:1,000- 1:10,000, immunocytochemistry: 1:500-1:2,000 | technique(s) immunohistochemistry: 1:50-1:200, immunoblotting: 0.04-0.4 μg/mL, immunofluorescence: 0.25-2 μg/mL | technique(s) western blot: 1:500-1:1000, ELISA: 1:10000 | technique(s) immunoprecipitation (IP): 2-10 μg/mg, western blot: 1:2,000-1:10,000 |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



