Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
100 μL | Check Cart for Availability | $540.00 |
About This Item
$540.00
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
affinity purified immunoglobulin
antibody product type
primary antibodies
clone
polyclonal
species reactivity
mouse, human
technique(s)
immunoprecipitation (IP): 2-10 μg/mg, proximity ligation assay: 10 μg, western blot: 1:2,000- 1:10,000
accession no.
NP_006302.2
shipped in
wet ice
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
rabbit ... NCoR(9611)
Immunogen
Physical form
Other Notes
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| description - | description Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody | description Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution | description Powered by Bethyl Laboratories, Inc. |
| species reactivity mouse, human | species reactivity human | species reactivity human | species reactivity mouse, human |
| antibody form affinity purified immunoglobulin | antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form affinity purified immunoglobulin |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate - |
| biological source rabbit | biological source rabbit | biological source rabbit | biological source rabbit |
| clone polyclonal | clone polyclonal | clone polyclonal | clone - |
| storage temp. 2-8°C | storage temp. −20°C | storage temp. −20°C | storage temp. 2-8°C |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



