Skip to Content
MilliporeSigma

Skip To

PLA0240

Rabbit anti-FANCE Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

FACE, FAE, Fanconi anemia, Protein FACE, complementation group E

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μL
Check Cart for Availability
$540.00

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IP, WB
Citations:
-

$540.00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

immunoprecipitation (IP): 5-10 μg/mg, western blot: 1:2,000- 1:10,000

accession no.

NP_068741.1

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... FANCE(2178)

Immunogen

The epitope recognized by PLA0240 maps to a region between residue 200 and 250 of human Fanconi anemia, complementation group E using the numbering given in entry NP_068741.1 (GeneID 2178).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

FANCE (Fanconi anemia, complementation group E) is a protein involved in DNA repair. Defects in FANCE are the cause of Fanconi anemia, a heterogenous autosomal recessive disorder characterized by congenital malformations and a predisposition to cancer. The FANCE gene is one of 13 complementation groups that include FANCA, FANCB, FANCC, FANCD1/BRCA2, FANCD2, FANCE, FANCF, FANCG, FANCI, FANCJ/BRIP1, FANCL, FANCM, and FANCN/PALB2. FANCE has been shown to mediate the interaction between FANCC and FANCD2.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library