Skip to Content
MilliporeSigma

Key Documents

Skip To

PRS2337

Anti-IKAP antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-IKBKAP, Anti-IKK complex-associated protein, Anti-Inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase complex-associated protein

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
$617.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
MDL number:
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ELISA (i), ICC, IF, WB
Citations:
-

$617.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 134-147 kDa

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable, immunofluorescence: suitable, indirect ELISA: suitable, western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IKBKAP(8518)

General description

IKAP (IKK-complex-associated protein) is considered as a subunit of the RNA Pol II elongator complex. It is also known as IKBKAP (Ikappakinase complex-associated protein or human elongator factor 1) and ELP1 (elongator protein-1). It has 37 exons and codes for 1332 amino acid IKAP protein, that is of ~150KD. It is present in most of the tissues. It is mapped to chromosome 9q31.

Immunogen

a 16 amino acid peptide from near the carboxy-terminus of human IKAP.

Biochem/physiol Actions

Mutations in IKAP cause familial dysautonomia. It helps to conserve the proliferation of neural precursor and blocks its unexpected differentiation into neurons. It plays an important role in cell motility and migration.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Other Notes

The action of this antibody can be blocked using blocking peptide SBP2337.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HPA049677HPA050686WH0008518M3
description

affinity isolated antibody, buffered aqueous solution

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

description

clone 6G9, purified immunoglobulin, buffered aqueous solution

species reactivity

mouse, human

species reactivity

human

species reactivity

human

species reactivity

human

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

6G9, monoclonal

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

mouse

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

storage temp.

−20°C

storage temp.

−20°C

storage temp.

-

storage temp.

-


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

WGK

WGK 2

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library