RAB1113
Properties
rabbit
ELISA: suitable
inter-assay cv: <12%intra-assay cv: <10%sensitivity: 6 pg/ml
wet ice
−20°C
Description
Show Differences
1 of 1
Sigma-Aldrich
Rabbit PDGF-BB ELISA Kit
RAB1152
Rat PDGF-BB ELISA Kit
RAB1107
Rabbit IL-17A ELISA Kit
RAB1109
Rabbit IL-2 ELISA Kit
for cell culture supernatants, plasma, and serum samples
rat, human
inter-assay cv: <12%intra-assay cv: <10%sensitivity: 8 pg/ml
inter-assay cv: <12%intra-assay cv: <10%sensitivity: 0.2 ng/ml
inter-assay cv: <12%intra-assay cv: <10%sensitivity: 2.4 pg/ml
Warning
Met. Corr. 1
10 - Combustible liquids
Not applicable
Choose from one of the most recent versions:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.