Skip to Content
MilliporeSigma

Skip To

RAB1287

Human BLAME ELISA

for serum, plasma and cell culture supernatants

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 kit
Check Cart for Availability
$979.00

About This Item

NACRES:
NA.32
UNSPSC Code:
41116158
Gene information:
human ... SLAMF8(56833)
Input:
sample type serum
sample type plasma
sample type cell culture supernatant(s)
Species reactivity:
human
Storage temp.:
−20°C
Shipped in:
wet ice

$979.00


Check Cart for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

human

technique(s)

ELISA: suitable

input

sample type serum
sample type plasma
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... SLAMF8(56833)

General description

This ELISA antibody pair detects Human BLAME (SLAMF8)

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
RAB1667RAB1343RAB1346
description

for serum, plasma and cell culture supernatants

description

for serum, plasma and cell culture supernatants

description

for serum, plasma and cell culture supernatants

description

for serum, plasma and cell culture supernatants

Gene Information

human ... SLAMF8(56833)

Gene Information

human ... RGMB(285704)

Gene Information

human ... CD48(962)

Gene Information

human ... CD58(965)

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

human

assay range

inter-assay cv: <12%
intra-assay cv: <10%

assay range

inter-assay cv: <12%
intra-assay cv: <10%

assay range

inter-assay cv: <12%
intra-assay cv: <10%

assay range

inter-assay cv: <12%
intra-assay cv: <10%

input

sample type serum
sample type plasma
sample type cell culture supernatant(s)

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum

input

sample type serum
sample type plasma
sample type cell culture supernatant(s)

input

sample type cell culture supernatant(s)
sample type plasma
sample type serum

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Pictograms

Corrosion

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class

10 - Combustible liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters