Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 kit | Check Cart for Availability | $979.00 |
About This Item
sample type cell culture supernatant(s)
sample type plasma
$979.00
species reactivity
human
technique(s)
ELISA: suitable
input
sample type serum
sample type cell culture supernatant(s)
sample type plasma
assay range
inter-assay cv: <12%
intra-assay cv: <10%
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... SLITRK1(114798)
Application
Please refer to the attached Protocolfor details.
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| description for serum, plasma and cell culture supernatants | description Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution | description Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution | description - |
| Gene Information human ... SLITRK1(114798) | Gene Information human ... SLITRK1(114798) | Gene Information human ... SLITRK1(114798) | Gene Information mouse ... Xiap(11798) |
| species reactivity human | species reactivity human | species reactivity human | species reactivity mouse |
| input sample type serum | input - | input - | input - |
| assay range inter-assay cv: <12% | assay range - | assay range - | assay range inter-assay cv: <10% |
| technique(s) ELISA: suitable | technique(s) immunofluorescence: 0.25-2 μg/mL (ICC-IF), immunohistochemistry: 1:200 - 1:500 | technique(s) immunohistochemistry: 1:200- 1:500 | technique(s) ELISA: suitable |
| shipped in wet ice | shipped in wet ice | shipped in wet ice | shipped in wet ice |
signalword
Warning
hcodes
pcodes
Hazard Classifications
Met. Corr. 1
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




