Skip to Content
MilliporeSigma

Skip To

RAB1767

Human UCH-L3 ELISA

for serum, plasma and cell culture supernatants

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 kit
Check Cart for Availability
$979.00

About This Item

NACRES:
NA.32
UNSPSC Code:
41116158
Gene information:
human ... UCHL3(7347)
Input:
sample type plasma
sample type cell culture supernatant(s)
sample type serum
Species reactivity:
human
Storage temp.:
−20°C
Shipped in:
wet ice

$979.00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

human

technique(s)

ELISA: suitable

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... UCHL3(7347)

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HPA019678RAB1766SAB2501667
description

for serum, plasma and cell culture supernatants

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

description

for serum, plasma and cell culture supernatants

description

affinity isolated antibody, buffered aqueous solution

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

rat, human, bovine, canine, pig

Gene Information

human ... UCHL3(7347)

Gene Information

human ... UCHL3(7347)

Gene Information

human ... UCHL1(7345)

Gene Information

human ... UCHL1(7345)

assay range

inter-assay cv: <12%
intra-assay cv: <10%

assay range

-

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.82 ng/mL
standard curve range: 0.819-200 ng/mL

assay range

-

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum

input

-

input

sample type plasma
sample type serum
sample type cell culture supernatant(s)

input

-

technique(s)

ELISA: suitable

technique(s)

immunoblotting: 0.04-0.4 μg/mL, immunofluorescence: 0.25-2 μg/mL, immunohistochemistry: 1:200-1:500

technique(s)

ELISA: suitable

technique(s)

immunohistochemistry: suitable, indirect ELISA: suitable, western blot: suitable

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters