Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
100 μL | Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery) | $617.00 |
About This Item
$617.00
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
26 kDa
species reactivity
horse, dog, mouse, bovine, human, rat, rabbit, guinea pig
concentration
0.5 mg - 1 mg/mL
technique(s)
western blot: suitable
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... TMEM127(55654)
Immunogen
Biochem/physiol Actions
Physical form
Other Notes
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| description affinity isolated antibody | description Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution | description IgG fraction of antiserum, buffered aqueous solution | description Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form IgG fraction of antiserum | antibody form affinity isolated antibody |
| biological source rabbit | biological source rabbit | biological source mouse | biological source rabbit |
| mol wt 26 kDa | mol wt - | mol wt - | mol wt - |
| shipped in wet ice | shipped in wet ice | shipped in dry ice | shipped in wet ice |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
10 - Combustible liquids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



