Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
100 μL | Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery) | $580.00 |
About This Item
$580.00
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
65kDa
species reactivity
human, guinea pig, rat, mouse
concentration
0.5 mg - 1 mg/mL
technique(s)
immunoblotting: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... CPEB2(132864)
Immunogen
Biochem/physiol Actions
Physical form
Other Notes
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| description affinity isolated antibody | description affinity isolated antibody | description purified immunoglobulin, buffered aqueous solution | description Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form purified immunoglobulin | antibody form affinity isolated antibody |
| biological source rabbit | biological source rabbit | biological source rabbit | biological source rabbit |
| species reactivity human, guinea pig, rat, mouse | species reactivity rat, human, mouse | species reactivity human | species reactivity human |
| Gene Information human ... CPEB2(132864) | Gene Information human ... ATF2(1386) | Gene Information human ... CPB2(1361) | Gene Information human ... CPB2(1361) |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
10 - Combustible liquids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



