Skip to Content
MilliporeSigma

Skip To

SAB3501166

Monoclonal Anti-Seasonal H1N1 Neuraminidase [10C5E11] antibody produced in mouse

purified antibody (Protein A)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Check Cart for Availability
$491.00

About This Item

NACRES:
NA.43
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
monoclonal
Application:
ELISA
Citations:
-

$491.00


Check Cart for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified antibody (Protein A)

antibody product type

primary antibodies

clone

monoclonal

form

liquid

species reactivity

Influenza A virus subtype H1N1

concentration

1 mg/mL

technique(s)

ELISA: suitable

isotype

IgG2b

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Seasonal H1N1 Neuraminidase Monoclonal Antibody: Influenza A virus has one of sixteen possible hemagglutinin (HA) surface proteins and one of nine possible neuraminidase (NA) surface proteins. In early 2009, a novel H1N1 swine-origin influenza (S-OIV) A virus was identified in specimens obtained from patients in Mexico and the United States. The genetic make-up of this swine flu virus is unlike any other: it is an H1N1 strain that combines a triple assortment first identified in 1998 including human, swine, and avian influenza with two new pig H3N2 virus genes from Eurasia, themselves of recent human origin.

Immunogen

Mouse monoclonal neuraminidase antibody was raised against a synthetic peptide from the seasonal influenza (H1N1) neuraminidase protein.

Application

Neuraminidase antibody can be used for the detection of neuraminidase protein from seasonal influenza A (H1N1) in ELISA. It will also detect 50 ng of free peptide at 1 μg/mL.

Physical form

Supplied in PBS containing 0.02% sodium azide.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB3501167SAB3501251SAB3501250
description

purified antibody (Protein A)

description

purified antibody (Protein A)

description

affinity isolated antibody

description

affinity isolated antibody

species reactivity

Influenza A virus subtype H1N1

species reactivity

Influenza A virus subtype H1N1

species reactivity

Influenza A virus subtype H1N1

species reactivity

Influenza A virus subtype H1N1

biological source

mouse

biological source

mouse

biological source

rabbit

biological source

rabbit

antibody form

purified antibody (Protein A)

antibody form

purified antibody (Protein A)

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

clone

monoclonal

clone

monoclonal

clone

polyclonal

clone

polyclonal

UniProt accession no.

B1AGS8

UniProt accession no.

B1AGS8

UniProt accession no.

B1AGS8

UniProt accession no.

B1AGS8, C4AL28


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

WGK

nwg

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters