Skip to Content
MilliporeSigma

Skip To

SAB5200345

Monoclonal Anti-Copper Transporting Atpase 1 - Percp antibody produced in mouse

clone S60-4, purified immunoglobulin

Synonym(s):

Anti- MC1, Anti-ATP7A, Anti-DSMAX, Anti-FLJ17790, Anti-SMAX3

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
$627.00

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
Peridinin-Chlorophyll-Protein Complex
Clone:
S60-4, monoclonal
Application:
IHC, IP, WB
Citations:
-

$627.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

Peridinin-Chlorophyll-Protein Complex

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

S60-4, monoclonal

form

buffered aqueous solution

mol wt

antigen predicted mol wt 180 kDa

species reactivity

mouse, human, rat

concentration

1 mg/mL

technique(s)

immunohistochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

isotype

IgG2b

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ATP7A(538)

Immunogen

Synthetic peptide amino acids 42-61 (cytoplasmic C-terminus) of human Copper- transporting ATPase1

Biochem/physiol Actions

Detects ~180kDa in rat brain membrane preparations

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

PBS pH7.4, 50% glycerol, 0.09% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB5200340SAB5200334SAB5200331
description

clone S60-4, purified immunoglobulin

description

clone S60-4, purified immunoglobulin

description

clone S60-4, purified immunoglobulin

description

clone S60-4, purified immunoglobulin

conjugate

Peridinin-Chlorophyll-Protein Complex

conjugate

Allophycocyanin conjugate

conjugate

Atto 594 conjugate

conjugate

Atto 390 conjugate

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

Gene Information

human ... ATP7A(538)

Gene Information

human ... ATP7A(538)

Gene Information

human ... ATP7A(538)

Gene Information

human ... ATP7A(538)

biological source

mouse

biological source

mouse

biological source

mouse

biological source

mouse

species reactivity

mouse, human, rat

species reactivity

rat, mouse, human

species reactivity

rat, mouse, human

species reactivity

human, mouse, rat

clone

S60-4, monoclonal

clone

S60-4, monoclonal

clone

S60-4, monoclonal

clone

S60-4, monoclonal


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters