Skip to Content
MilliporeSigma

Skip To

SCP0258

Urotensin I carp

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
500 μg
Usually ships in 2 weeks.
$307.00

About This Item

Empirical Formula (Hill Notation):
C208H336N64O67S2
Molecular Weight:
4869.41
NACRES:
NA.32
UNSPSC Code:
12352200
Form:
lyophilized
Assay:
≥95% (HPLC)

$307.00


Usually ships in 2 weeks.

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥72%

storage condition

protect from light

storage temp.

−20°C

Application

Urotensin-I is a peptide ligand, corticotrophin-releasing factor (CRF) receptor, used to study its immunoregulatory activities and behavorial effects in fish.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SCP0178SCP0132SCP0236
description

-

description

-

description

-

description

-

assay

≥95% (HPLC)

assay

≥95% (HPLC)

assay

≥95% (HPLC)

assay

≥95% (HPLC)

form

lyophilized

form

lyophilized

form

lyophilized

form

lyophilized

storage condition

protect from light

storage condition

protect from light

storage condition

protect from light

storage condition

protect from light

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

composition

Peptide Content, ≥72%

composition

Peptide Content, ≥73%

composition

Peptide Content, ≥77%

composition

Peptide Content, ≥75%


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters