Skip to Content
MilliporeSigma

Skip To

SML1033

A-278637

≥98%

Synonym(s):

(9S)-9-(3-Bromo-4-fluorophenyl)-2,3,5,6,7,9-hexahydro-thieno[3,2-b]quinolin-8(4H)-one 1,1-dioxide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 mg
Check Cart for Availability
$269.00

About This Item

Empirical Formula (Hill Notation):
C17H15BrFNO3S
CAS Number:
Molecular Weight:
412.27
NACRES:
NA.77
UNSPSC Code:
12352202
Assay:
≥98%
Form:
powder

$269.00


Check Cart for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98%

form

powder

optical activity

[α]/D -44 to -54°, c = 0.1 in acetonitrile

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

Biochem/physiol Actions

A-278637 is a selective opener of ATP-sensitive K (KATP) channels (EC50 = 102 nM) that blocks contraction of bladder smooth muscle in both in vivo and in vitro assays. A-278637 is 15-fold more selective than nifedipine for bladder compared other smooth muscle KATP channels, and displays reduced effects on mean arterial pressure compared to other channel openers.
A-278637 is a selective opener of ATP-sensitive K (KATP) channels.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML1640SML0813SML1392
description

≥98%

description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder (flocculent)

assay

≥98%

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 5 mg/mL, clear (warmed)

solubility

DMSO: 10 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear (warmed)

solubility

DMSO: 10 mg/mL, clear

color

white to beige

color

white to light brown

color

white to beige

color

white to beige


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters