Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
1 mg | Check Cart for Availability | $324.00 |
About This Item
NACRES:
NA.77
UNSPSC Code:
51111800
Assay:
≥95% (mixture of Leucinostatin A+B, HPLC)
Form:
powder
$324.00
Check Cart for Availability
Quality Segment
biological source
(Paecilomyces lilacinus)
assay
≥95% (mixture of Leucinostatin A+B, HPLC)
form
powder
solubility
DMF: 10 mg/mL, DMSO: 10 mg/mL, ethanol: 10 mg/mL, methanol: 10 mg/mL
storage temp.
−20°C
Biochem/physiol Actions
Leucinostatins A and B are the major components of an atypical nonapeptide complex produced by Paecilomyces lilacinus.
Leucinostatins A and B are the major components of an atypical nonapeptide complex produced by Paecilomyces lilacinus. Leucinostatins display broad bio-activity against gram-positive bacteria, fungi, plants and tumor cell lines. They play a role as uncouplers in the inner mitochondria membrane and inhibit mitochondrial ATP synthesis. Recently, interest in Leucinostatins has focused on other aspects such as understanding their activity: insulin-like growth factor I regulators, ionophore, inhibitors of cell surface expression of viral glycoproteins and their potent in vitro and in vivo anti-trypanosomal activity.
Preparation Note
Mixture of Leucinostatin A (Cas# 76600-38-9 / MF: C62H111N11O13) and Leucinostatin B (Cas# 76663-52-0 / MF: C61H109N11O13).
Other Notes
Mixture of Leucinostatin A (Cas# 76600-38-9 / MF: C62H111N11O13) and Leucinostatin B (Cas# 76663-52-0 / MF: C61H109N11O13).
Stable at −20 °C for at least 2 years.
1 of 1
This Item | |||
|---|---|---|---|
| description ≥95% (mixture of Leucinostatin A+B, HPLC), (Paecilomyces lilacinus) | description European Pharmacopoeia (EP) Reference Standard | description European Pharmacopoeia (EP) Reference Standard | description United States Pharmacopeia (USP) Reference Standard |
| assay ≥95% (mixture of Leucinostatin A+B, HPLC) | assay - | assay - | assay - |
| form powder | form - | form - | form - |
| Quality Level 200 | Quality Level - | Quality Level - | Quality Level - |
| storage temp. −20°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. −20°C |
| solubility methanol: 10 mg/mL, DMF: 10 mg/mL, DMSO: 10 mg/mL | solubility - | solubility - | solubility - |
| biological source (Paecilomyces lilacinus) | biological source - | biological source - | biological source - |
Analyte
Description
- Leuconostatin A
- Leucinostatin B
Storage Class
11 - Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



