Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
5 mg | Estimated to ship onOctober 05, 2026fromMILWAUKEE | $69.30 |
About This Item
Empirical Formula (Hill Notation):
C19H20N2O5S
CAS Number:
Molecular Weight:
388.44
UNSPSC Code:
12352202
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Quality Segment
assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 20 mg/mL, clear
storage temp.
2-8°C
SMILES string
O=C(C1=C(N2CCOCC2)C=CC([N+]([O-])=O)=C1)OCC3=CC=C(SC)C=C3
InChI
1S/C19H20N2O5S/c1-27-16-5-2-14(3-6-16)13-26-19(22)17-12-15(21(23)24)4-7-18(17)20-8-10-25-11-9-20/h2-7,12H,8-11,13H2,1H3
InChI key
VPXGOSNPXCNGAI-UHFFFAOYSA-N
Biochem/physiol Actions
IMB5046 is a microtubule inhibitor that has antitumor activity, overcoming multidrug-resistance.
IMB5046 is a microtubule inhibitor that has antitumor activity, overcoming multidrug-resistance. IMB5046 was cytotoxic with an IC50 range of 37 to 426 nM against multiple tumor cell lines, including some that were resistant to colchicine, vincristine and paclitaxel. IMB5046 was also active against human lung tumor xenografts. IMB5046 was found to bind to tubulin at the colchicine site, but with a different binding mode. IMB5046 inhibited tubulin polymerization, and induced G2/M arrest.
1 of 1
This Item | ||
|---|---|---|
| description ≥98% (HPLC) | description Pharmaceutical Secondary Standard; Certified Reference Material | description - |
| form powder | form - | form liquid |
| assay ≥98% (HPLC) | assay - | assay - |
| Quality Level 100 | Quality Level 300 | Quality Level 100 |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. −20°C |
| solubility DMSO: 20 mg/mL, clear | solubility - | solubility - |
| color white to beige | color - | color - |
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


