Skip to Content
MilliporeSigma

Skip To

SML2454

Q151

≥98% (HPLC)

Synonym(s):

(2S)-N-[2-(4-cyanophenyl)-5-benzoxazolyl]-2-[(2,3-difluorophenyl)amino]propanamide, (S)-N-(2-(4-cyanophenyl)benzo[d]oxazol-5-yl)-2-((2,3-difluorophenyl)amino)propanamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg

Available to ship TODAYfromMILWAUKEE

$94.40

About This Item

Empirical Formula (Hill Notation):
C23H16F2N4O2
CAS Number:
Molecular Weight:
418.40
UNSPSC Code:
51111800
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder

$94.40


Available to ship TODAYDetails


​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

FC1=C(N[C@@H](C)C(NC2=CC(N=C(C3=CC=C(C#N)C=C3)O4)=C4C=C2)=O)C=CC=C1F

InChI

1S/C23H16F2N4O2/c1-13(27-18-4-2-3-17(24)21(18)25)22(30)28-16-9-10-20-19(11-16)29-23(31-20)15-7-5-14(12-26)6-8-15/h2-11,13,27H,1H3,(H,28,30)/t13-/m0/s1

InChI key

QUSYLBPSSGPXBL-ZDUSSCGKSA-N

Biochem/physiol Actions

Inosine 5′-monophosphate dehydrogenase 2 (IMPDH2) inhibitor
Q151 is an inhibitor of bacterial inosine 5′-monophosphate dehydrogenase 2 (IMPDH2) with an IC50 value of 18 nM. It does not inhibit human IMPDH

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HPA001400WH0003615M1I1782
description

≥98% (HPLC)

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

description

clone 1E12-B6, purified immunoglobulin, buffered aqueous solution

description

recombinant, expressed in E. coli

form

powder

form

buffered aqueous glycerol solution

form

buffered aqueous solution

form

solution

assay

≥98% (HPLC)

assay

-

assay

-

assay

-

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−70°C

solubility

DMSO: 2 mg/mL, clear

solubility

-

solubility

-

solubility

-

color

white to beige

color

-

color

-

color

-

IMPDH2 inhibitor for enzyme inhibition studies

Anti-IMP dehydrogenase 2 antibody immunoblotting, Anti-Inosine-5′-monophosphate dehydrogenase 2 antibody, Anti-IMPDH2 antibody for immunohistochemistry, Anti-IMPDH2 antibody rabbit, rabbit Anti-IMPDH2 antibody, immunohistochemistry Anti-IMPDH2 antibody 1:500-1:1000, Anti-IMPDH2 antibody immunofluorescence grade, Anti-IMPDH2 rabbit polyclonal antibody, Anti-IMPDH-II antibody for western blotting, Anti-IMPDH2 Prestige Antibody Sigma-Aldrich, rabbit Anti-IMPDH2 antibody validated by HPA, Anti-IMPDH2 antibody for research, Anti-IMPDH2 antibody 0.04-0.4 µg/mL for immunoblotting

-

-


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters