Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
5 mg | Available to ship TODAYfromMILWAUKEE | $99.20 |
25 mg | Available to ship TODAYfromMILWAUKEE | $400.00 |
About This Item
Empirical Formula (Hill Notation):
C24H24N2O2S
CAS Number:
Molecular Weight:
404.52
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
−20°C
SMILES string
CC(C=C1)=CC=C1OCCN2C(SCCOC3=CC=CC=C3)=NC4=CC=CC=C42
InChI
1S/C24H24N2O2S/c1-19-11-13-21(14-12-19)27-16-15-26-23-10-6-5-9-22(23)25-24(26)29-18-17-28-20-7-3-2-4-8-20/h2-14H,15-18H2,1H3
InChI key
PNCBJXVNIRRHJP-UHFFFAOYSA-N
Biochem/physiol Actions
MEPB is a modulator of the activation function-2 (AF-2) of the androgen receptor (AR), increasing AR co-repressor binding. MEPB specifically binds the BF3 pocket of the androgen receptor where it modulates binding of co-regulators to the AF2 domain. MEPB was found to rescue degeneration in a mouse model of spinal bulbar muscular atrophy, a motor neuron disease caused by toxic gain of function of the androgen receptor.
Modulator of the activation function-2 (AF-2) of the androgen receptor (AR)
1 of 1
This Item | |||
|---|---|---|---|
| description ≥98% (HPLC) | description 97% | description PESTANAL®, analytical standard | description United States Pharmacopeia (USP) Reference Standard |
| form powder | form solid | form - | form - |
| assay ≥98% (HPLC) | assay 97% | assay - | assay - |
| storage temp. −20°C | storage temp. - | storage temp. - | storage temp. - |
| solubility DMSO: 2 mg/mL, clear | solubility - | solubility - | solubility - |
| color white to beige | color - | color - | color - |
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



