Skip to Content
MilliporeSigma

SML2661

A-108 Hydrochloride

≥98% (HPLC)

Synonym(s):

A-108 Hydrochloride, 6-[(2R)-2-(4-Cyanophenyl)-2-hydroxy-2-(1-methyl-1H-imidazol-5-yl)ethoxy]-4′-(trifluoromethoxy)[1,1′-biphenyl]-3-carbonitrile hydrochloride, A-409108 Hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C27H19F3N4O3 · HCl
Molecular Weight:
540.92
NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

A-108 (A-409108), an ABT-100 inactive enantiomer, serves as negative control for ABT-100.
negative control for ABT-100


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library