Skip to Content
MilliporeSigma

Skip To

SML2688

J30-8

≥98% (HPLC)

Synonym(s):

(5Z)-2-(2-Chlorophenylimino)-5-(5-fluoro-2-oxoindolin-3-ylidene)thiazolidin-4-one, 2-((2-Chlorophenyl)imino)-5-((Z)-5-fluoro-2-oxoindolin-3-ylidene)thiazolidin-4-one

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg

Estimated to ship TODAYfromMILWAUKEE

$86.00
$73.10
25 mg

Estimated to ship TODAYfromMILWAUKEE

$348.00
$295.80

About This Item

Empirical Formula (Hill Notation):
C17H9ClFN3O2S
CAS Number:
Molecular Weight:
373.79
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder

$73.10

List Price$86.00Save 15%
Web-Only Promotion

Estimated to ship TODAYDetails


​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

Fc1cc2c(cc1)NC(=O)\C\2=C3/SC(=NC/3=O)Nc4c(cccc4)Cl

InChI

1S/C17H9ClFN3O2S/c18-10-3-1-2-4-12(10)21-17-22-16(24)14(25-17)13-9-7-8(19)5-6-11(9)20-15(13)23/h1-7H,(H,20,23)(H,21,22,24)/b14-13-

InChI key

HNOFQKYBQYGFNQ-YPKPFQOOSA-N

Biochem/physiol Actions

J30-8 is a potent c-Jun N-terminal kinase JNK3 subtype-selective inhibtior (human JNK3 IC50 = 40 nM, JNK1α1 & JNK2α2 IC50 >100 μM) that exhibits excellent kinome-wide selectivity (IC50 >7.7 μM against 398 kinase targets) and neuroprotective efficacy against Aβ25-35 toxicity in SH-SY5Y cultures (1 μM). When compared with the pan-JNK inhibitor SP600125 in the APPswe/PS1dE9 murine AD model in vivo (30 mg/kg ip.), J30-8 offers superior therapeutic efficacy and pharmacokinetic properties with longer retention time (elimination t1/2 = 16.11 vs 1.23 hrs; plasma conc = 41 ng/mL vs 22 ng/mL 24 hr post ip.) and greater brain-permeability (brain/plasma ratio = 0.40 vs. 0.14 8 hr post ip.).
Potent JNK3 subtype-selective c-Jun N-terminal kinase inhibtior with superior in vivo efficacy, pharmacokinetics and brain-permeability in AD mice than SP600125.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML0176SML1839SML1725
description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: ≥15 mg/mL

solubility

DMSO: 15 mg/mL, clear

solubility

DMSO: 20 mg/mL, clear

color

white to beige

color

light yellow to dark yellow

color

yellow to orange

color

yellow to orange


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters