Skip to Content
MilliporeSigma

Skip To

SML2698

EVT-101 dihyrochloride

≥98% (HPLC)

Synonym(s):

5-(3-Difluoromethyl-4-fluorophenyl)-3-(2-methylimidazol-1-ylmethyl)pyridazine dihydrochloride, 5-[3-(Difluoromethyl)-4-fluorophenyl]-3-[(2-methyl-1H-imidazol-1-yl)methyl]pyridazine dihydrochloride, EVT 101 dihydrochloride, EVT101 dihydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg

Available to ship TODAYfromMILWAUKEE

$130.00
25 mg

Available to ship TODAYfromMILWAUKEE

$423.00

About This Item

Empirical Formula (Hill Notation):
C16H13F3N4 · 2HCl
CAS Number:
Molecular Weight:
391.22
NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated

$130.00


Available to ship TODAYDetails


​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC(F)C1=C(F)C=CC(C2=CC(CN3C(C)=NC=C3)=NN=C2)=C1

InChI

1S/C16H13F3N4.2ClH/c1-10-20-4-5-23(10)9-13-6-12(8-21-22-13)11-2-3-15(17)14(7-11)16(18)19;;/h2-8,16H,9H2,1H3;2*1H

InChI key

OJBLXSPBJMGZDN-UHFFFAOYSA-N

Biochem/physiol Actions

EVT-101 is a potent and orally active NR2B-subtype selective N-methyl-D-aspartate (NMDA) receptor antagonist (IC50 = 17 nM against 5 nM Ro 25-6981 for binding rat brain membrane) that targets the GluN1/GluN2B dimer interface with a distinctly different binding mode as ifenprodil. EVT 101 potently inhibits current induction by Glu/Gly (100 μM each) in rat GluN1-1a/mouse GluN2B co-expressing xenopus in vitro (IC50 = 12 nM) and blocks MK-801 binding in mouse brain in vivo (3 mg EVT-101/kg p.o. & 0.2 μCi [3H]MK-801/g i.v. administered 55 min and 10 min prior to brain removal, respectively).
Potent and orally active NR2B-/GluN2B-subtype selective N-methyl-D-aspartate (NMDA) receptor antagonist with in vitro and in vivo efficacy.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML4355791423HPA041915
description

≥98% (HPLC)

description

≥95% (HPLC)

description

-

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

form

powder

form

oil

form

powder (maroon)

form

buffered aqueous glycerol solution

assay

≥98% (HPLC)

assay

≥95% (HPLC)

assay

-

assay

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-

storage temp.

−20°C

storage condition

desiccated

storage condition

-

storage condition

-

storage condition

-

solubility

H2O: 2 mg/mL, clear

solubility

-

solubility

-

solubility

-

color

white to beige

color

colorless to yellow

color

-

color

-


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters