Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
5 mg | Available to ship TODAYfromMILWAUKEE | $73.10 |
25 mg | Available to ship TODAYfromMILWAUKEE | $298.00 |
About This Item
Empirical Formula (Hill Notation):
C17H18FN3S
CAS Number:
Molecular Weight:
315.41
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Quality Segment
assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
2-8°C
SMILES string
Fc1cc(c(cc1)N3CCCCC3)NC(=S)c2ccncc2
InChI key
VRKZHYSJZOUICG-UHFFFAOYSA-N
Biochem/physiol Actions
FIT-039 is a potent and selective cyclin-dependent kinase 9 (CDK9) inhibitor that binds to the ATP -binding pocket of CDK9 and inhibits the kinase activity of P-TEFb complex. FIT-039 inhibits replication of herpes simplex virus 1 (HSV-1), HSV-2, human adenovirus, and human cytomegalovirus in cultured cells. FIT-039 ointment inhibits formation of skin legion in a mice HSV-1 infection model.
potent and selective cyclin-dependent kinase 9 (CDK9) inhibitor; antiviral agent
1 of 1
This Item | |||
|---|---|---|---|
| description ≥98% (HPLC) | description ≥98% (HPLC) | description clone LP14, from mouse | description clone 10A3, Chemicon®, from mouse |
| assay ≥98% (HPLC) | assay ≥98% (HPLC) | assay - | assay - |
| form powder | form powder | form - | form - |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 300 |
| storage temp. 2-8°C | storage temp. −20°C | storage temp. - | storage temp. - |
| solubility DMSO: 2 mg/mL, clear | solubility DMSO: 2 mg/mL, clear | solubility - | solubility - |
| color white to beige | color faint yellow to dark orange | color - | color - |
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



