Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
5 mg | Check Cart for Availability | $70.60 $60.01 |
25 mg | Check Cart for Availability | $288.00 $244.80 |
About This Item
Empirical Formula (Hill Notation):
C20H18N2O3
CAS Number:
Molecular Weight:
334.37
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Quality Segment
assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
2-8°C
SMILES string
N(CC)(CC)c1cc2[o][c](c(cc2cc1)c3nc4c([o]3)cccc4)=O
InChI
1S/C20H18N2O3/c1-3-22(4-2)14-10-9-13-11-15(20(23)25-18(13)12-14)19-21-16-7-5-6-8-17(16)24-19/h5-12H,3-4H2,1-2H3
InChI key
UJOQSHCJYVRZKJ-UHFFFAOYSA-N
Biochem/physiol Actions
EMI1 (ChemBridge 5213777) is a call penetrant and potent inhibitor of EGFR signaling in EGFR L858R/T790M/C797S and ex19del/T790M/C797S triple mutants associated with drug-resistant NSCLC. Additionally, EMI1 potently depolymerized interphase microtubules, disrupts spindle formation and induced mitotic block in PC9 EGFR ex19del/T790M/C797S cells. EMI1 exhibits potent anti-proliferative activity against various cancer cell lines.
call penetrant and potent inhibitor of EGFR signaling in EGFR L858R/T790M/C797S triple mutants associated with drug-resistant NSCLC
1 of 1
This Item | |||
|---|---|---|---|
| description ≥98% (HPLC) | description ≥97% (HPLC) | description ≥98% (HPLC) | description ≥98% (HPLC) |
| form powder | form powder | form powder | form powder |
| assay ≥98% (HPLC) | assay ≥97% (HPLC) | assay ≥98% (HPLC) | assay ≥98% (HPLC) |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| storage temp. 2-8°C | storage temp. room temp | storage temp. room temp | storage temp. 2-8°C |
| solubility DMSO: 2 mg/mL, clear | solubility DMSO: ≥5 mg/mL | solubility DMSO: ≥25 mg/mL | solubility DMSO: 2 mg/mL, clear |
| color white to beige | color white to beige | color white to tan | color white to beige |
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



