Skip to Content
MilliporeSigma

Skip To

SML3016

Afobazole

≥98% (HPLC)

Synonym(s):

2-((2-Morpholino)ethylthio)-5-ethoxybenzimidazole, 5-Ethoxy-2-[[2-(4-morpholinyl)ethyl]thio]-1H-benzimidazole, 6-Ethoxy-2-[[2-(4-morpholinyl)ethyl]thio]-1H-benzimidazole, Afobazol, Aphobazole, CM 346, Fabomotizole

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg

Available to ship TODAYfromMILWAUKEE

$83.10
25 mg

Available to ship TODAYfromMILWAUKEE

$338.00

About This Item

Empirical Formula (Hill Notation):
C15H21N3O2S
CAS Number:
Molecular Weight:
307.41
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
film

$83.10


Available to ship TODAYDetails


Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

film

color

white to beige

storage temp.

2-8°C

SMILES string

CCOC1=CC2=C(C=C1)N=C(SCCN3CCOCC3)N2

InChI

1S/C15H21N3O2S/c1-2-20-12-3-4-13-14(11-12)17-15(16-13)21-10-7-18-5-8-19-9-6-18/h3-4,11H,2,5-10H2,1H3,(H,16,17)

InChI key

WWNUCVSRRUDYPP-UHFFFAOYSA-N

Biochem/physiol Actions

Afobazole is an anxiolytic that appears to act via σ-1 and σ2 receptors. Afobazole exhibit neuroprotective potential in both in vivo and in vitro models. It is a potent modulator of microglia.
anxiolytic that appears to act via σ-1 and σ-2 receptors

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML2825I5409SML1133
description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC), solid

description

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

form

film

form

powder

form

solid

form

powder

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

color

white to beige

color

white to beige

color

off-white to light yellow

color

white to beige


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters