Skip to Content
MilliporeSigma

Skip To

SML3052

ppTG21 trifluoroacetate salt

≥95% (HPLC)

Synonym(s):

GLFHALLHLLHSLWHLLLHA-OH TFA salt, Gly-Leu-Phe-His-Ala-Leu-Leu-His-Leu-Leu-His-Ser-Leu-Trp-His-Leu-Leu-Leu-His-Ala TFA salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mg

Available to ship TODAYfromMILWAUKEE

$129.00

About This Item

Empirical Formula (Hill Notation):
C115H173N31O22 · xC2HF3O2
Molecular Weight:
2341.80 (free base basis)
NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥95% (HPLC)
Form:
film or powder

$129.00


Available to ship TODAYDetails


Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

film or powder

color

white to beige

storage temp.

−20°C

Biochem/physiol Actions

Endosomolytic agent that facilitates cell type-specific gene editing by enabling Cas9 RNP endosome escape following receptor-mediated endocytosis.
ppTG21 is an endosomolytic agent (pI 7.7, charge +1.3 @ pH 7.4) that facilitates cell type-specific gene editing by enabling endosome escape of Cas9 ribonucleoprotein (RNP) complex following its uptake via receptor-mediated endocytosis. Following liver-specific asialoglycoprotein receptor (ASGPr)-mediated delivery of ASGPr ligand (ASGPrL)-harboring & SV40 nuclear localization sequence (NLS)-bearing S. pyogenes Cas9 (Cas9-2lig-1NLS) RNP with EMX1-targeting sgRNA, target gene editing occurs only upon coincubation of the RNP in the presence of 30 molar equiv of ppTG21 (ave 4.8% indels in HepG2 cells vs. 0.2% indels without ppTG21).

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML3264SML2626SML2891
description

≥95% (HPLC)

description

≥95% (HPLC)

description

≥95% (HPLC)

description

≥95% (HPLC)

assay

≥95% (HPLC)

assay

≥95% (HPLC)

assay

≥95% (HPLC)

assay

≥95% (HPLC)

form

film or powder

form

powder

form

film

form

lyophilized powder

Quality Level

100

Quality Level

100

Quality Level

-

Quality Level

100

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

color

white to beige

color

white to off-white

color

white to beige

color

white to off-white


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. What is the recommended solvent to dissolve Product No. SML3052?

    1 answer
    1. This product can be dissolved in sterile distilled water.

      Helpful?

Reviews

No rating value

Active Filters