Skip to Content
MilliporeSigma

Skip To

SML3113

trans-22a

≥98% (HPLC)

Synonym(s):

1-[trans-4-(3-Methylphenyl)cyclohexyl]-4-(3-pyridinyl)piperazine, trifluoroacetate (1:2), trans-1-(4-(3-Methylphenyl)cyclohexyl)-4-((pyridin-4-yl)methyl)piperazine, trifluoroacetate (1:2)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg

Available to ship TODAYfromMILWAUKEE

$129.00
25 mg

Available to ship TODAYfromMILWAUKEE

$518.00

About This Item

Empirical Formula (Hill Notation):
C22H29N3 · 2CF3CO2H
CAS Number:
Molecular Weight:
563.53
NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder

$129.00


Available to ship TODAYDetails


Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

Inactive trans-isomer of and negative control for the potent and highly selective TrpV6 (CaT1, ECaC2) inhibitor cis-22a.
trans-22a is the inactive trans-isomer of and negative control for the potent and highly selective TrpV6 (CaT1, ECaC2) inhibitor cis-22a (Cd2+ influx IC50 = 3.13 μM/trans-22a vs. 320 nM/cis-22a using hTRPV6 transfected HEK293).

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML3112SML3156SML1205
description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

H2O: 2 mg/mL, clear

solubility

H2O: 2 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 20 mg/mL, clear

color

white to beige

color

white to beige

color

white to beige

color

white to light brown


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters